Learn More
Biomatik Corporation AMA-1 anti-Plasmodium falciparum, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507046
Encompass_Preferred
The AMA-1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Plasmodium falciparum. Immunogen: Recombinant Plasmodium falciparum Apical membrane antigen 1 protein (25-546AA). Applications: ELISA. Alternative Names: Merozoite surface antigen, AMA-1. UniProt: P22621. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7046 Supplier Biomatik Corporation Supplier No. CAC07460
The AMA-1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Plasmodium falciparum. Immunogen: Recombinant Plasmodium falciparum Apical membrane antigen 1 protein (25-546AA). Applications: ELISA. Alternative Names: Merozoite surface antigen, AMA-1. UniProt: P22621. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.