Learn More
Biomatik Corporation FAM46C anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507062
Encompass_Preferred
The FAM46C Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Protein FAM46C protein (1-391AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Protein FAM46C, FAM46C. UniProt: Q5VWP2. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7062 Supplier Biomatik Corporation Supplier No. CAC07476
The FAM46C Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Protein FAM46C protein (1-391AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Protein FAM46C, FAM46C. UniProt: Q5VWP2. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.