Learn More
Biomatik Corporation ATP5A1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07482
Encompass_Preferred
The ATP5A1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human ATP synthase subunit alpha, mitochondrial protein (44-553AA). Applications: ELISA, WB. Recommended dilution: WB: 1:500-2000. Alternative Names: ATP synthase subunit alpha, mitochondrial, ATP5A1, ATP5A, ATP5AL2, ATPM. UniProt: P25705. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7067