Learn More
Biomatik Corporation AURKB anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07483
Encompass_Preferred
The AURKB Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Aurora kinase B protein (1-255AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Aurora kinase B, Aurora 1, Aurora- and IPL1-like midbody-associated protein 1, AIM-1, Aurora/IPL1-related kinase 2, ARK-2, Aurora-related kinase 2, STK-1, Serine/threonine-protein kinase 12, Serine/threonine-protein kinase 5, Serine/threonine-protein kinase aurora-B, AURKB, AIK2, AIM1, AIRK2, ARK2, STK1, STK12, STK5. UniProt: Q96GD4. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7068