Learn More
Biomatik Corporation AZIN1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07484
Encompass_Preferred
The AZIN1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Antizyme inhibitor 1 protein (101-400AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Antizyme inhibitor 1, AZI, Ornithine decarboxylase antizyme inhibitor, AZIN1, OAZI, OAZIN. UniProt: O14977. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7069