Learn More
Biomatik Corporation STK24 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07485
Encompass_Preferred
Stk24 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to STK24. The Stk24 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Serine; Threonine-Protein Kinase 24 Protein (1-431Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: STK24. Accession Number: Q9Y6E0. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7070