Learn More
Biomatik Corporation BCL10 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507072
Encompass_Preferred
The BCL10 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human B-cell lymphoma/leukemia 10 protein (1-233AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Bcl-10, CARD-containing molecule enhancing NF-kappa-B, CARD-like apoptotic protein, hCLAP, CED-3/ICH-1 prodomain homologous E10-like regulator, CIPER, cCARMEN, Cellular-E10, c-E10, mE10, BCL10, CIPER, CLAP. UniProt: O95999. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7072 Supplier Biomatik Corporation Supplier No. CAC07487
The BCL10 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human B-cell lymphoma/leukemia 10 protein (1-233AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Bcl-10, CARD-containing molecule enhancing NF-kappa-B, CARD-like apoptotic protein, hCLAP, CED-3/ICH-1 prodomain homologous E10-like regulator, CIPER, cCARMEN, Cellular-E10, c-E10, mE10, BCL10, CIPER, CLAP. UniProt: O95999. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.