Learn More
Biomatik Corporation BRCC3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07488
Encompass_Preferred
The BRCC3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Lys-63-specific deubiquitinase BRCC36 protein (2-316AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: BRCA1-A complex subunit BRCC36 BRCA1/BRCA2-containing complex subunit 3 BRCA1/BRCA2-containing complex subunit 36 BRISC complex subunit BRCC36 BRCC3 BRCC36, C6.1A, CXorf53. UniProt: P46736. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7073