Learn More
Biomatik Corporation CAPN1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507075
Encompass_Preferred
The CAPN1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Calpain-1 catalytic subunit protein (1-714AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200. Alternative Names: Calcium-activated neutral proteinase 1, Calpain mu-type, Calpain-1 large subunit, Cell proliferation-inducing gene 30 protein, Micromolar-calpain, muCANP, CAPN1, CANPL1, PIG30. UniProt: P07384. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7075 Supplier Biomatik Corporation Supplier No. CAC07490
The CAPN1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Calpain-1 catalytic subunit protein (1-714AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200. Alternative Names: Calcium-activated neutral proteinase 1, Calpain mu-type, Calpain-1 large subunit, Cell proliferation-inducing gene 30 protein, Micromolar-calpain, muCANP, CAPN1, CANPL1, PIG30. UniProt: P07384. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.