Learn More
Biomatik Corporation CD2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507076
Encompass_Preferred
The CD2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human T-cell surface antigen CD2 protein (25-209AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Erythrocyte receptor, LFA-2, LFA-3 receptor, Rosette receptor, T-cell surface antigen T11/Leu-5, CD2, SRBC. UniProt: P06729. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7076 Supplier Biomatik Corporation Supplier No. CAC07491
The CD2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human T-cell surface antigen CD2 protein (25-209AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Erythrocyte receptor, LFA-2, LFA-3 receptor, Rosette receptor, T-cell surface antigen T11/Leu-5, CD2, SRBC. UniProt: P06729. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.