Learn More
Biomatik Corporation COPZ1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07495
Encompass_Preferred
The COPZ1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Coatomer subunit zeta-1 protein (1-177AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Coatomer subunit zeta-1, Zeta-1-coat protein, Zeta-1 COP, COPZ1, COPZ, CGI-120, HSPC181. UniProt: P61923. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7080