Learn More
Biomatik Corporation CYCS anti-Human, Mouse, Rat, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07497
Encompass_Preferred
The CYCS Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse, Rat. Immunogen: Recombinant Human Cytochrome c protein (2-105AA). Applications: ELISA, WB, IHC. Recommended dilution: WB:1:500-5000, IHC:1:200-1:500. Alternative Names: CYCS, CYC. UniProt: P99999. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7082