Learn More
Biomatik Corporation CYP17A1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507083
Encompass_Preferred
The CYP17A1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Steroid 17-alpha-hydroxylase/17,20 lyase protein (1-508AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: 17-alpha-hydroxyprogesterone aldolase, CYPXVII, Cytochrome P450 17A1, Cytochrome P450-C17, Cytochrome P450c17, Steroid 17-alpha-monooxygenase, CYP17A1. UniProt: P05093. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7083 Supplier Biomatik Corporation Supplier No. CAC07498
The CYP17A1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Steroid 17-alpha-hydroxylase/17,20 lyase protein (1-508AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: 17-alpha-hydroxyprogesterone aldolase, CYPXVII, Cytochrome P450 17A1, Cytochrome P450-C17, Cytochrome P450c17, Steroid 17-alpha-monooxygenase, CYP17A1. UniProt: P05093. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.