Learn More
Biomatik Corporation DHRS3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507086
Encompass_Preferred
The DHRS3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Short-chain dehydrogenase/reductase 3 protein (30-169AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Short-chain dehydrogenase/reductase 3, DD83.1, Retinal short-chain dehydrogenase/reductase 1, retSDR1, Retinol dehydrogenase 17, Short chain dehydrogenase/reductase family 16C member 1, DHRS3, RDH17, SDR16C1, UNQ2424/PRO4983. UniProt: O75911. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7086 Supplier Biomatik Corporation Supplier No. CAC07501
The DHRS3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Short-chain dehydrogenase/reductase 3 protein (30-169AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Short-chain dehydrogenase/reductase 3, DD83.1, Retinal short-chain dehydrogenase/reductase 1, retSDR1, Retinol dehydrogenase 17, Short chain dehydrogenase/reductase family 16C member 1, DHRS3, RDH17, SDR16C1, UNQ2424/PRO4983. UniProt: O75911. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.