Learn More
Biomatik Corporation DNALI1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07502
Encompass_Preferred
The DNALI1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Axonemal dynein light intermediate polypeptide 1 protein (1-258AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Axonemal dynein light intermediate polypeptide 1, Inner dynein arm light chain, axonemal, hp28, DNALI1. UniProt: O14645. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7087