Learn More
Biomatik Corporation Acta1 anti-Mouse, Rat, Zebrafish, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07504
Encompass_Preferred
The Acta1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse, Rat, Zebrafish. Immunogen: Mouse skeletal muscle alpha Actin (Native Protein). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. UniProt: P68134. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7089