Learn More
Biomatik Corporation Eno1 anti-Mouse, Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07505
Encompass_Preferred
The Eno1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse, Human. Immunogen: Recombinant mouse Alpha-enolase protein (2-433AA). Applications: ELISA, WB. Recommended dilution: WB: 1:1000-1:5000. Alternative Names: 2-phospho-D-glycerate hydro-lyase, Enolase 1, Non-neural enolase, Eno1, Eno-1. UniProt: P17182. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7090