Learn More
Biomatik Corporation GDI2 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507095
Encompass_Preferred
The GDI2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Rab GDP dissociation inhibitor beta protein (1-445AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-5000, IHC:1:20-1:200. Alternative Names: Rab GDP dissociation inhibitor beta, Rab GDI beta, Guanosine diphosphate dissociation inhibitor 2, GDI-2, GDI2, RABGDIB. UniProt: P50395. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7095 Supplier Biomatik Corporation Supplier No. CAC07510
The GDI2 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Rab GDP dissociation inhibitor beta protein (1-445AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-5000, IHC:1:20-1:200. Alternative Names: Rab GDP dissociation inhibitor beta, Rab GDI beta, Guanosine diphosphate dissociation inhibitor 2, GDI-2, GDI2, RABGDIB. UniProt: P50395. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.