Learn More
Biomatik Corporation IFNA14 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07511
Encompass_Preferred
Ifna14 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IFNA14. The Ifna14 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Interferon Alpha-14 Protein (24-189Aa). Applications: ELISA, WB, IHC. Target Synonyms: IFNA14. Accession Number: P01570. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7096