Learn More
Biomatik Corporation DYNLL1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07514
Encompass_Preferred
The DYNLL1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Dynein light chain 1, cytoplasmic protein (1-89AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: 8 kDa dynein light chain??DLC8??Dynein light chain LC8-type 1??Protein inhibitor of neuronal nitric oxide synthase??PIN. UniProt: P63167. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7099