Learn More
Biomatik Corporation PTAFR anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507104
Encompass_Preferred
Ptafr Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PTAFR. The Ptafr Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Platelet-Activating Factor Receptor Protein (297-342Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PTAFR. Accession Number: P25105. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7104 Supplier Biomatik Corporation Supplier No. CAC07519
Ptafr Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PTAFR. The Ptafr Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Platelet-Activating Factor Receptor Protein (297-342Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: PTAFR. Accession Number: P25105. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.