Learn More
Biomatik Corporation CSF3 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07534
Encompass_Preferred
The CSF3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Granulocyte colony-stimulating factor (27-200AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: Pluripoietin, INN=Filgrastim, INN=Lenograstim, CSF3, C17orf33, GCSF. UniProt: P09919. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7119