Learn More
Biomatik Corporation GC anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507121
Encompass_Preferred
The GC Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Vitamin D-binding protein (19-474AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Gc-globulin, Group-specific component, GC. UniProt: P02774. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7121 Supplier Biomatik Corporation Supplier No. CAC07536
The GC Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Vitamin D-binding protein (19-474AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200. Alternative Names: Gc-globulin, Group-specific component, GC. UniProt: P02774. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.