Learn More
Biomatik Corporation Ccl4 anti-Mouse, Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07539
Encompass_Preferred
The Ccl4 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Mouse, Human. Immunogen: Recombinant mouse C-C motif chemokine 4 protein (24-92AA). Applications: ELISA, IHC. Recommended dilution: IHC:1:20-1:200. Alternative Names: C-C motif chemokine 4, Immune activation protein 2, ACT-2, ACT2, Macrophage inflammatory protein 1-beta, MIP-1-beta, Protein H400, SIS-gamma, Small-inducible cytokine A4, Ccl4, Mip1b, Scya4. UniProt: P14097. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7124