Learn More
Biomatik Corporation MUC16 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507126
Encompass_Preferred
Muc16 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MUC16. The Muc16 Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Mucin-16 Protein (14067-14328Aa). Applications: ELISA, IHC. Target Synonyms: MUC16. Accession Number: Q8WXI7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7126 Supplier Biomatik Corporation Supplier No. CAC07541
Muc16 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MUC16. The Muc16 Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Mucin-16 Protein (14067-14328Aa). Applications: ELISA, IHC. Target Synonyms: MUC16. Accession Number: Q8WXI7. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.