Learn More
Biomatik Corporation SHBG anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07543
Encompass_Preferred
Shbg Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to SHBG. The Shbg Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Sex Hormone-Binding Globulin Protein (30-402Aa). Applications: ELISA, WB. Target Synonyms: SHBG. Accession Number: P04278. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7128