Learn More
Biomatik Corporation Ren2 anti-Mouse, Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07555
Encompass_Preferred
Ren2 Polyclonal Antibody is a Mus musculus (Mouse (Mus musculus)) Polyclonal Antibody to Ren2. The Ren2 Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Mus musculus (Mouse (Mus musculus)). Immunogen: Recombinant Mouse Renin-2 Protein (64-351Aa). Applications: ELISA, WB. Target Synonyms: Ren2. Accession Number: P00796. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7140