Learn More
Biomatik Corporation EHHADH anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07561
Encompass_Preferred
The EHHADH Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Peroxisomal bifunctional enzyme protein (1-723AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200. Alternative Names: PBE, PBFE, EHHADH, ECHD. UniProt: Q08426. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7146