Learn More
Biomatik Corporation ELAVL1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507149
Encompass_Preferred
The ELAVL1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human ELAV-like protein 1 protein (2-326AA). Applications: ELISA, WB. Recommended dilution: WB: 1:500-2000. Alternative Names: ELAV-like protein 1, ELAVL1, HUR, Hu-antigen R, HuR. UniProt: Q15717. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7149 Supplier Biomatik Corporation Supplier No. CAC07564
The ELAVL1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human ELAV-like protein 1 protein (2-326AA). Applications: ELISA, WB. Recommended dilution: WB: 1:500-2000. Alternative Names: ELAV-like protein 1, ELAVL1, HUR, Hu-antigen R, HuR. UniProt: Q15717. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.