Learn More
Biomatik Corporation EXOSC5 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507161
Encompass_Preferred
The EXOSC5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Exosome complex component RRP46 protein (1-235AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200. Alternative Names: Exosome complex component RRP46, EXOSC5, CML28, RRP46, Chronic myelogenous leukemia tumor antigen 28, Exosome component 5, Ribosomal RNA-processing protein 46, p12B. UniProt: Q9NQT4. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7161 Supplier Biomatik Corporation Supplier No. CAC07576
The EXOSC5 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human. Immunogen: Recombinant human Exosome complex component RRP46 protein (1-235AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:500-2000, IHC:1:20-1:200. Alternative Names: Exosome complex component RRP46, EXOSC5, CML28, RRP46, Chronic myelogenous leukemia tumor antigen 28, Exosome component 5, Ribosomal RNA-processing protein 46, p12B. UniProt: Q9NQT4. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.