Learn More
Biomatik Corporation MCM9 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07580
Encompass_Preferred
Mcm9 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MCM9. The Mcm9 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Dna Helicase Mcm9 Protein (1-391Aa). Applications: ELISA, WB, IHC. Target Synonyms: MCM9. Accession Number: Q9NXL9. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7165