Learn More
Biomatik Corporation NUP58 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507166
Encompass_Preferred
Nup58 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NUP58. The Nup58 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Nucleoporin P58; P45 Protein (211-480Aa). Applications: ELISA, WB, IHC. Target Synonyms: NUP58. Accession Number: Q9BVL2. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7166 Supplier Biomatik Corporation Supplier No. CAC07581
Nup58 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to NUP58. The Nup58 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Nucleoporin P58; P45 Protein (211-480Aa). Applications: ELISA, WB, IHC. Target Synonyms: NUP58. Accession Number: Q9BVL2. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.