Learn More
Biomatik Corporation TRMT6 anti-Human, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507169
Encompass_Preferred
Trmt6 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TRMT6. The Trmt6 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Trna (Adenine (58) -N (1) ) -Methyltransferase Non-Catalytic Subunit Trm6 Protein (1-300Aa). Applications: ELISA, WB, IHC. Target Synonyms: TRMT6. Accession Number: Q9UJA5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7169 Supplier Biomatik Corporation Supplier No. CAC07584
Trmt6 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TRMT6. The Trmt6 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Trna (Adenine (58) -N (1) ) -Methyltransferase Non-Catalytic Subunit Trm6 Protein (1-300Aa). Applications: ELISA, WB, IHC. Target Synonyms: TRMT6. Accession Number: Q9UJA5. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.