Learn More
Biomatik Corporation COLEC10 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07586
Encompass_Preferred
The COLEC10 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Collectin-10 protein (28-277AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:200-1000, IHC:1:20-1:200. Alternative Names: Collectin liver protein 1, CL-L1, Collectin-34, CL-34, UNQ366/PRO702, CLL1, COLEC10. UniProt: Q9Y6Z7. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7171