Learn More
Biomatik Corporation FIS1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07594
Encompass_Preferred
The FIS1 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Mitochondrial fission 1 protein (1-122AA). Applications: ELISA, WB, IHC, IF, IP. Recommended dilution: WB:1:1000-1:5000, IHC:1:500-1:1000, IF:1:50-1:500, IP:1:200-1:2000. Alternative Names: Mitochondrial fission 1 protein, FIS1 homolog, hFis1, Tetratricopeptide repeat protein 11, TPR repeat protein 11, FIS1, TTC11, CGI-135. UniProt: Q9Y3D6. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7179