Learn More
Biomatik Corporation GSTM3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07595
Encompass_Preferred
The GSTM3 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Glutathione S-transferase Mu 3 protein (1-225AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Glutathione S-transferase Mu 3, GST class-mu 3, GSTM3-3, hGSTM3-3, GSTM3, GST5. UniProt: P21266. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7180