Learn More
Biomatik Corporation WISP1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507183
Encompass_Preferred
Wisp1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to WISP1. The Wisp1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Wnt1-Inducible-Signaling Pathway Protein 1 Protein (130-251Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: WISP1. Accession Number: O95388. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7183 Supplier Biomatik Corporation Supplier No. CAC07598
Wisp1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to WISP1. The Wisp1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Wnt1-Inducible-Signaling Pathway Protein 1 Protein (130-251Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: WISP1. Accession Number: O95388. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.