Learn More
Biomatik Corporation CPT1A anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07604
Encompass_Preferred
The CPT1A Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Carnitine O-palmitoyltransferase 1, liver isoform protein (301-600AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:2000-1:5000, IHC:1:20-1:200. Alternative Names: Carnitine O-palmitoyltransferase 1, liver isoform, Carnitine O-palmitoyltransferase I, liver isoform, CPT I, CPTI-L, Carnitine palmitoyltransferase 1A, CPT1A, CPT1. UniProt: P50416. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7189