Learn More
Biomatik Corporation FCER1A anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07608
Encompass_Preferred
The FCER1A Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human High affinity immunoglobulin epsilon receptor subunit alpha protein (26-205AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:2000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: High affinity immunoglobulin epsilon receptor subunit alpha, Fc-epsilon RI-alpha, FcERI, IgE Fc receptor subunit alpha, FCER1A, FCE1A. UniProt: P12319. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7193