Learn More
Biomatik Corporation GAA anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07609
Encompass_Preferred
The GAA Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Lysosomal alpha-glucosidase protein (601-952AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:1000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Lysosomal alpha-glucosidase, Acid maltase, Aglucosidase alfa, 76 kDa lysosomal alpha-glucosidase, 70 kDa lyso, GAA. UniProt: P10253. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7194