Learn More
Biomatik Corporation HOXD9 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07610
Encompass_Preferred
The HOXD9 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Homeobox protein Hox-D9 protein (38-276AA). Applications: ELISA, WB, IHC. Recommended dilution: WB: 1:2000-1:10000, IHC:1:20-1:200. Alternative Names: Homeobox protein Hox-D9, Homeobox protein Hox-4C, Homeobox protein Hox-5.2, HOXD9, HOX4C. UniProt: P28356. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7195