Learn More
Biomatik Corporation MEIS1 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07616
Encompass_Preferred
Meis1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to MEIS1. The Meis1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Homeobox Protein Meis1 Protein (1-90Aa). Applications: ELISA, WB, IHC, IP. Target Synonyms: MEIS1. Accession Number: O00470. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7201