Learn More
Biomatik Corporation PRG2 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07618
Encompass_Preferred
Prg2 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to PRG2. The Prg2 Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Bone Marrow Proteoglycan Protein (151-195Aa). Applications: ELISA, IHC. Target Synonyms: PRG2. Accession Number: P13727. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7203