Learn More
Biomatik Corporation RHD anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07623
Encompass_Preferred
Rhd Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to RHD. The Rhd Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Blood Group Rh (D) Polypeptide Protein (32-76Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: RHD. Accession Number: Q02161. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7208