Learn More
Biomatik Corporation TMEM64 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07629
Encompass_Preferred
Tmem64 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TMEM64. The Tmem64 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Transmembrane Protein 64 Protein (40-118Aa). Applications: ELISA, WB, IHC. Target Synonyms: TMEM64. Accession Number: Q6YI46. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7214