Learn More
Biomatik Corporation TOX3 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07630
Encompass_Preferred
Tox3 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to TOX3. The Tox3 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Tox High Mobility Group Box Family Member 3 Protein (8-305Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: TOX3. Accession Number: O15405. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7215