Learn More
Biomatik Corporation WRB anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Catalog No. 501507220
Encompass_Preferred
Wrb Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to WRB. The Wrb Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Tail-Anchored Protein Insertion Receptor Wrb Protein (38-99Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: WRB. Accession Number: O00258. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7220 Supplier Biomatik Corporation Supplier No. CAC07635
Wrb Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to WRB. The Wrb Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Tail-Anchored Protein Insertion Receptor Wrb Protein (38-99Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: WRB. Accession Number: O00258. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.