Learn More
Biomatik Corporation YWHAG anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07636
Encompass_Preferred
Ywhag Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to YWHAG. The Ywhag Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human 14-3-3 Protein Gamma Protein (113-194Aa). Applications: ELISA, WB, IHC, IP. Target Synonyms: YWHAG. Accession Number: P61981. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7221