Learn More
Biomatik Corporation CD34 anti-Human, Mouse, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07639
Encompass_Preferred
The CD34 Polyclonal Antibody is raised in Rabbit. Species Reactivity: Human, Mouse. Immunogen: Recombinant human Hematopoietic progenitor cell antigen CD34 protein (124-375AA). Applications: ELISA, WB, IHC, IF. Recommended dilution: WB: 1:2000-1:5000, IHC:1:20-1:200, IF:1:50-1:200. Alternative Names: Hematopoietic progenitor cell antigen CD34, CD34, CD34. UniProt: P28906. Purification: >95%, Protein G purified. Storage: -20C. Restriction: For Research Use Only.
Catalog No. 50-150-7224