Learn More
Biomatik Corporation IKZF1 anti-Human, Rabbit Polyclonal Antibody, 100UL
Supplier:  Biomatik Corporation CAC07645
Encompass_Preferred
Ikzf1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to IKZF1. The Ikzf1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Recombinant Human Dna-Binding Protein Ikaros Protein (1-265Aa). Applications: ELISA, WB, IHC, IF. Target Synonyms: IKZF1. Accession Number: Q13422. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-7230